8-Bromoisoquinoline

Supplier: Cayman Chemical

10007082-1 10007082-500
CAYM10007082-1EA 113 EUR
CAYM10007082-1 CAYM10007082-500
8-Bromoisoquinoline
8-Bromoisoquinoline
Formula: C₉H₆BrN
MDL Number: MFCD04973298
CAS Number: 63927-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR