Triethyl phosphonoacetate 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate
139701000
139705000
139700025
ACRO139701000EA
59.9
EUR
ACRO139701000
ACRO139705000
ACRO139700025
Triethyl phosphonoacetate 97%
Triethyl phosphonoacetate
Formula:
C₈H₁₇O₅P MW: 224,19 g/mol Boiling Pt: 260…262 °C (760 mmHg) Melting Pt: –24 °C Density: 1,13 g/cm³ Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009177 CAS Number: 867-13-0 EINECS: 212-757-6 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4300 to 1.4320 (20°C, 589 nm) |
GC | ≥96.0 % |
Color scale | ≤50 APHA |
Acid value | ≤2 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...