Ethyl (ethoxymethylene)cyanoacetate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Ethyl 2-Cyano-3-Ethoxyacrylate
A12366.0B
A12366.18
A12366.30
A12366.0BEA
378
EUR
A12366.0B
A12366.18
A12366.30
Ethyl (ethoxymethylene)cyanoacetate ≥98%
Ethyl (ethoxymethylene)cyanoacetate
Purity: 98%
CAS No.: 94-05-3
Molecular Formula: C8H11NO3
Formula Weight: 169.18
Flash Point: 130°(266°F)
Melting Point: 47-52°
Boiling Point: 190-191°/30mm
Beilstein Reference: 878814
UN No.: 3439
MDL No.: MFCD00009136
Formula:
C₈H₁₁NO₃ MW: 169,18 g/mol Boiling Pt: 190…191 °C (30 mmHg) Melting Pt: 47…52 °C Flash Pt: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009136 CAS Number: 94-05-3 EINECS: 202-299-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...