Ammonium carbamate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Carbamic acid ammonium salt
18134.18
18134.30
18134.18EA
54
EUR
18134.18
18134.30
Ammonium carbamate ≥98%
Ammonium carbamate
Purity: 98%
CAS No.: 1111-78-0
Molecular Formula: CH6N2O2
Formula Weight: 78.07
Physical Form: Powder
Density: 1.6
Appearance: Off-white
Melting Point: 60° subl.
Solubility: Freely soluble in water. Soluble in alcohol
Beilstein Reference: 3914091
Merck Reference: 14,507
MDL No.: MFCD00013010
Ammoniating agent
Formula:
H₂NCO₂NH₄ MW: 78,07 g/mol Melting Pt: 60 °C (subl.) Density: 1,6 Storage Temperature: Ambient |
MDL Number:
MFCD00013010 CAS Number: 1111-78-0 EINECS: 214-185-2 Merck Index: 14,507 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...