Phenyl carbamate ≥98%
Supplier: Thermo Scientific
L00944.06
L00944.14
L00944.06EA
34.7
EUR
L00944.06
L00944.14
Phenyl carbamate ≥98%
Phenyl carbamate
Purity: 98+%
CAS No.: 622-46-8
Molecular Formula: H2NCO2C6H5
Formula Weight: 137.14
Melting Point: 149-152°
Beilstein Reference: 2042627
Formula:
C₇H₇NO₂ MW: 137,14 g/mol Melting Pt: 149…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007961 CAS Number: 622-46-8 EINECS: 210-737-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...