N,N,N',N',N''-Pentamethyldiethylenetriamine for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine

M-Clarity™ quality level = MQ200

8.41692.0100 8.41692.0500
8.41692.0100EA 115 EUR
8.41692.0100 8.41692.0500
N,N,N',N',N''-Pentamethyldiethylenetriamine for synthesis, Sigma-Aldrich®
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formula: C₉H₂₃N₃
MW: 173,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014876
CAS Number: 3030-47-5
EINECS: 221-201-1
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Solubility (20 °C) miscible
Molar Mass 173.3 g/mol
Boiling Point 199 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 77 °C
Explosion Limit 1.1 - 5.7 %(V)
Density 0.828 g/cm3 (20 °C)
pH 11.9 (100 g/l, H2O, 20 °C)
Ignition Point 155 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR