2-(Allyloxy)benzaldehyde ≥97%

Supplier: Thermo Fisher Scientific
Warning

B25119.06 B25119.14
B25119.06EA 0 EUR
B25119.06 B25119.14
2-(Allyloxy)benzaldehyde ≥97%
2-(Allyloxy)benzaldehyde

Purity: 97% CAS No.: 28752-82-1 Molecular Formula: C10H10O2 Formula Weight: 162.19 Flash Point: 110°(230°F) Refractive Index: 1.5500 Boiling Point: 129-132°/10mm MDL No.: MFCD00014130

Formula: C₁₀H₁₀O₂
Boiling Pt: 129…132 °C (10 mmHg)
Density: 1,079
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014130
CAS Number: 28752-82-1
EINECS: 249-198-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR