4-Benzyloxy-2-methoxybenzaldehyde ≥98%
Supplier: Thermo Scientific
Synonyms:
4-(Benzyloxy)-2-methoxybenzaldehyde
L15516.03
L15516.06
L15516.03EA
0
EUR
L15516.03
L15516.06
4-Benzyloxy-2-methoxybenzaldehyde ≥98%
4-Benzyloxy-2-methoxybenzaldehyde
Purity: 98+%
CAS No.: 58026-14-5
Molecular Formula: C15H14O3
Formula Weight: 242.27
Melting Point: 93-98°
Boiling Point: ca 180°/2mm
MDL No.: MFCD01075699
Formula:
C₁₅H₁₄O₃ MW: 242,2700 g/mol Melting Pt: 93…98 °C |
MDL Number:
MFCD01075699 CAS Number: 58026-14-5 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...