Potassium dicyanoargentate (54,2% Ag) ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

12551.14 12551.22 12551.36
12551.14EA 122 EUR
12551.14 12551.22 12551.36
Potassium dicyanoargentate (54,2% Ag) ≥99.9% (metals basis)
Potassium dicyanoargentate

Purity: 99.9% CAS No.: 506-61-6 Molecular Formula: C2AgKN2 Formula Weight: 199.01 Physical Form: Powder Density: 2.36 Appearance: White Solubility: Soluble in water Merck Reference: 14,7669 UN No.: 1588 MDL No.: MFCD00036297


Silver plating

Formula: C₂AgKN₂
Density: 2,36
Storage Temperature: Ambient
MDL Number: MFCD00036297
CAS Number: 506-61-6
EINECS: 208-047-0
UN: 1588
ADR: 6.1,I
Merck Index: 14,7669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR