1-Chloroisoquinoline 95%
Supplier: Thermo Scientific
|
Warning
|
L17831.03
L17831.06
L17831.03EA
38.7
EUR
L17831.03
L17831.06
1-Chloroisoquinoline 95%
1-Chloroisoquinoline
Purity: 95%
CAS No.: 19493-44-8
Molecular Formula: C9H6ClN
Formula Weight: 163.61
Flash Point:
Formula:
C₉H₆ClN MW: 163,61 g/mol Boiling Pt: 105…108 °C (4 mmHg) Melting Pt: 33…35 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00024134 CAS Number: 19493-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...