5-Chloro-3-methylbenzo[b]thiophene ≥97%
Supplier: Thermo Scientific
A11827.14
A11827.14EA
645
EUR
A11827.14
5-Chloro-3-methylbenzo[b]thiophene ≥97%
5-Chloro-3-methylbenzo[b]thiophene
Purity: 97%
CAS No.: 19404-18-3
Molecular Formula: C9H7ClS
Formula Weight: 182.67
Melting Point: 33-36°
MDL No.: MFCD00052506
Formula:
C₉H₇ClS MW: 182,67 g/mol Melting Pt: 33…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052506 CAS Number: 19404-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...