(S)-(-)-Indoline-2-carboxylic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-Indoline-2-carboxylic acid
H27009.03
H27009.06
H27009.03EA
54.8
EUR
H27009.03
H27009.06
(S)-(-)-Indoline-2-carboxylic acid ≥97%
(S)-(-)-Indoline-2-carboxylic acid
Purity: 97+%
CAS No.: 79815-20-6
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: ca 177° dec.
Optical Rotation: -115° (c=1 in 1N HCl)
Formula:
C₉H₉NO₂ MW: 163,18 g/mol Melting Pt: ∼ 177 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00070578 CAS Number: 79815-20-6 EINECS: 410-860-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...