2-Thienylacetyl chloride 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride
232230500
232230050
ACRO232230500EA
177
EUR
ACRO232230500
ACRO232230050
2-Thienylacetyl chloride 99%
2-Thienylacetyl chloride
Formula:
C₆H₅ClOS MW: 160,62 g/mol Boiling Pt: 130…135 °C (90 mmHg) Flash Pt: 101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005456 CAS Number: 39098-97-0 EINECS: 254-290-0 UN: 1760 ADR: 8,III |
Specification Test Results
Appearance | Clear brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5500 to 1.5520 (20°C, 589 nm) |
GC | ≥98.5 % |
Free acid | ≤2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...