3-Thienylacetic acid 98%

Supplier: Thermo Fisher Scientific
Warning

205370100 205370500
ACRO205370100EA 108 EUR
ACRO205370100 ACRO205370500
3-Thienylacetic acid 98%
3-Thienylacetic acid
Formula: C₆H₆O₂S
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00005473
CAS Number: 6964-21-2
EINECS: 230-166-1
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to beige Powder or flakes
Infrared spectrum Conforms
Melting point 77 to 80 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR