S-Isopropylisothiouronium bromide

Supplier: Cayman Chemical

81290-100 81290-50 81290-500
CAYM81290-100EA 98.9 EUR
CAYM81290-100 CAYM81290-50 CAYM81290-500
S-Isopropylisothiouronium bromide
S-Isopropylisothiouronium bromide
Formula: C₄H₁₁BrN₂S
CAS Number: 57200-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR