Ampicillin sodium salt

Supplier: Molekula
Danger

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

23337642-100G 23337642-25G
MOLE23337642-100GEA 528 EUR
MOLE23337642-100G MOLE23337642-25G
Ampicillin sodium salt
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371,39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR