2'-Deoxyuridine

Supplier: Molekula

61613969-100G 61613969-5G
MOLE61613969-100GEA 531 EUR
MOLE61613969-100G MOLE61613969-5G
2'-Deoxyuridine
2'-Deoxyuridine
Formula: C₉H₁₂N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00006527
CAS Number: 951-78-0
EINECS: 213-455-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR