Glycylglycine (Gly-Gly)

Supplier: Roth Carl

3794.1 3794.2 3794.3 3794.4
ROTH3794.1EA 0 EUR
ROTH3794.1 ROTH3794.2 ROTH3794.3 ROTH3794.4
Glycylglycine (Gly-Gly)
Proteins and Peptides

  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR