Cytidine

Supplier: Roth Carl

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

0717.1 0717.2
ROTH0717.1EA 63.8 EUR
ROTH0717.1 ROTH0717.2
Cytidine
Cytidine
Formula: C₉H₁₃N₃O₅
MW: 243,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006545
CAS Number: 65-46-3
EINECS: 200-610-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR