Didodecyldimethylammonium bromide ≥98%

Supplier: Thermo Scientific
Warning

B22839.04 B22839.09
B22839.04EA 38.2 EUR
B22839.04 B22839.09
Didodecyldimethylammonium bromide ≥98%
Didodecyldimethylammonium bromide

Purity: 98+% CAS No.: 3282-73-3 Molecular Formula: C26H56BrN Formula Weight: 462.65 Melting Point: 157-162° Beilstein Reference: 3576662

Formula: C₂₆H₅₆BrN
MW: 462,65 g/mol
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00041969
CAS Number: 3282-73-3
EINECS: 221-923-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR