2,4'-Bipyridyl ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2,4-Bipyridine, 2,4'-Bipyridine
H27366.MD
H27366.03
H27366.MDEA
102
EUR
H27366.MD
H27366.03
2,4'-Bipyridyl ≥97%
2,4'-Bipyridyl
Purity: 97%
CAS No.: 581-47-5
Molecular Formula: C10H8N2
Formula Weight: 156.19
Melting Point: 59-63°
Boiling Point: 281°
UN No.: 2811
MDL No.: MFCD00006217
Formula:
C₁₀H₈N₂ Boiling Pt: 281 °C Melting Pt: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006217 CAS Number: 581-47-5 EINECS: 209-467-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...