2-Chloro-N-methylacetamide ≥97%
Supplier: Thermo Scientific
|
Warning
|
H32362.06
H32362.14
H32362.06EA
81.8
EUR
H32362.06
H32362.14
2-Chloro-N-methylacetamide ≥97%
2-Chloro-N-methylacetamide
Purity: 97%
CAS No.: 96-30-0
Molecular Formula: C3H6ClNO
Formula Weight: 107.54
Melting Point: 45-47°
MDL No.: MFCD00018913
Formula:
C₃H₆ClNO MW: 107,54 g/mol Melting Pt: 45…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018913 CAS Number: 96-30-0 EINECS: 202-497-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...