Alizarin red S (sodium salt)

Supplier: Roth Carl
Warning

Synonyms: Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium

0348.1 0348.2
ROTH0348.1EA 31.7 EUR
ROTH0348.1 ROTH0348.2
Alizarin red S (sodium salt)
Alizarin Red S (sodium salt)
Formula: C₁₄H₇NaO₇S
MW: 342,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3
EINECS: 204-981-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR