(+)-Dibenzoyl-D-tartaric acid, anhydrous 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid
B24754.09
B24754.18
B24754.30
B24754.09EA
25.8
EUR
B24754.09
B24754.18
B24754.30
(+)-Dibenzoyl-D-tartaric acid, anhydrous 99%
(+)-Dibenzoyl-D-tartaric acid
Purity: 99%
CAS No.: 17026-42-5
Molecular Formula: C18H14O8
Formula Weight: 358.31
Melting Point: 154-156°
Beilstein Reference: 2227343
Formula:
C₁₈H₁₄O₈ MW: 358,31 g/mol Melting Pt: 154…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063222 CAS Number: 17026-42-5 EINECS: 241-097-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...