2-Amino-6-iodopurine 97%
Supplier: Thermo Scientific
Synonyms:
6-Iodo-9H-purin-2-amine
H51672.03
H51672.06
H51672.03EA
0
EUR
H51672.03
H51672.06
2-Amino-6-iodopurine 97%
2-Amino-6-iodopurine
Purity: 97%
CAS No.: 19690-23-4
Molecular Formula: C5H4IN5
Formula Weight: 261.02
MDL No.: MFCD08706375
Formula:
C₅H₄IN₅ MW: 261,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08706375 CAS Number: 19690-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...