(±)-Tetrahydrofurfuryl bromide 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(Bromomethyl)tetrahydrofuran

A15192.06 A15192.09 A15192.18
A15192.06EA 30.6 EUR
A15192.06 A15192.09 A15192.18
(±)-Tetrahydrofurfuryl bromide 95%
(±)-Tetrahydrofurfuryl bromide

Purity: 95% CAS No.: 1192-30-9 Molecular Formula: C5H9BrO Formula Weight: 165.04 Storage Temperature: 2°C to 8°C Flash Point: 61°(142°F) Refractive Index: 1.4880 Boiling Point: 169-171° Beilstein Reference: 102717 MDL No.: MFCD00016894

Formula: C₅H₉BrO
Boiling Pt: 169…171 °C
Density: 1,452
Flash Pt: 61 °C (142 °F)
MDL Number: MFCD00016894
CAS Number: 1192-30-9
EINECS: 214-750-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR