Methyl green zinc chloride salt, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: CI 42590, Ethyl Green, Methyl green zinc chloride double salt, Methyl Green

M-Clarity™ quality level = MQ200

198080-10G 198080-50G
198080-10GEA 241 EUR
198080-10G 198080-50G
Methyl green zinc chloride salt, Sigma-Aldrich®
Methyl Green zinc chloride salt
Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS Number: 7114-03-6
EINECS: 230-415-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7022caee36158e3bdb25b3e8bf856f0822f67c5d&vid=d2cd36b4936c8aa3b530803ae0c1b1354a453497&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR