N,N-Dimethylmethanesulphonamide ≥98%
Supplier: Thermo Fisher Scientific
A14454.06
A14454.14
A14454.06EA
34.2
EUR
A14454.06
A14454.14
N,N-Dimethylmethanesulphonamide ≥98%
N,N-Dimethylmethanesulphonamide
Purity: 98+%
CAS No.: 918-05-8
Molecular Formula: CH3SO2N(CH3)2
Formula Weight: 123.17
Melting Point: 46-50°
Boiling Point: 119-120°/17mm
MDL No.: MFCD00014740
Formula:
C₃H₉NO₂S Boiling Pt: 119…120 °C (17 mmHg) Melting Pt: 46…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014740 CAS Number: 918-05-8 EINECS: 213-039-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...