(4-Methoxyphenyl)acetyl chloride ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Methoxyphenylacetyl chloride

L15256.06 L15256.14
L15256.06EA 147 EUR
L15256.06 L15256.14
(4-Methoxyphenyl)acetyl chloride ≥98%
(4-Methoxyphenyl)acetyl chloride

Purity: 98% CAS No.: 4693-91-8 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Flash Point: 110°(230°F) Refractive Index: 1.5400 Boiling Point: 143°/10mm Beilstein Reference: 908061 UN No.: 3265 MDL No.: MFCD00075474

Formula: C₉H₉ClO₂
MW: 184,62 g/mol
Boiling Pt: 143 °C (10 mmHg)
Density: 1,208
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075474
CAS Number: 4693-91-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR