MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt)
Supplier: Roth Carl
Synonyms:
Sodium 2-hydroxy-3-morpholinopropane-1-sulphonate
7101.1
7101.2
ROTH7101.1EA
102
EUR
ROTH7101.1
ROTH7101.2
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulphonic acid sodium salt)
MOPSO sodium salt (3-(4-Morpholino)-2-hydroxypropanesulphonic acid sodium salt)
Formula:
C₇H₁₄NNaO₅S MW: 247,2445 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070007 CAS Number: 79803-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...