3-Quinolineboronic acid 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
L20088.03
L20088.06
L20088.MD
L20088.03EA
49.5
EUR
L20088.03
L20088.06
L20088.MD
3-Quinolineboronic acid 95%
3-Quinolineboronic acid
Purity: 95%
CAS No.: 191162-39-7
Molecular Formula: C9H8BNO2
Formula Weight: 172.98
Melting Point:
Formula:
C₉H₈BNO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD02183527 CAS Number: 191162-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...