Niobium(V) ethanolate 99.9% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Niobium(V) ethoxide
L16031.03
L16031.06
L16031.03EA
59.2
EUR
L16031.03
L16031.06
Niobium(V) ethanolate 99.9% (metals basis)
Niobium(V) ethanolate
Purity: 99.9%
CAS No.: 3236-82-6
Molecular Formula: Nb(OC2H5)5
Formula Weight: 318.21
Flash Point: 36°(97°F)
Refractive Index: 1.5160
Melting Point: 6°
Boiling Point: 140-142°/0.1mm
Beilstein Reference: 3678883
UN No.: 2924
MDL No.: MFCD00015122
Formula:
C₁₀H₂₅NbO₅ MW: 318.209 g/mol Boiling Pt: 140…142 °C (0.1 mmHg) Melting Pt: 6 °C Density: 1.258 Flash Pt: 36 °C (97 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015122 CAS Number: 3236-82-6 EINECS: 221-795-2 UN: 2924 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...