Chromotrope 2B ≥97%

Supplier: Thermo Scientific

L11330.14 L11330.22
L11330.14EA 63 EUR
L11330.14 L11330.22
Chromotrope 2B ≥97%
Chromotrope 2B

CAS No.: 548-80-1 Molecular Formula: C16H9N3Na2O10S2 Formula Weight: 513.37 Melting Point: 300° Beilstein Reference: 4109733 Merck Reference: 14,2240 MDL No.: MFCD00003951

Formula: C₁₆H₉N₃Na₂O₁₀S₂
MW: 513,37 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003951
CAS Number: 548-80-1
EINECS: 208-959-9
Merck Index: 14,2240

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR