2'-Deoxyguanosine hydrate 99+%
Supplier: Thermo Scientific
Synonyms:
9-(2-Deoxy-β-D-ribofuranosyl)guanine hydrate, Guanine-2′-deoxyriboside hydrate, 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one hydrate(1:x)
226731000
226730010
226730050
ACRO226731000EA
45
EUR
ACRO226731000
ACRO226730010
ACRO226730050
2'-Deoxyguanosine hydrate 99+%
2'-Deoxyguanosine hydrate
Formula:
C₁₀H₁₃N₅O₄·xH₂O MW: 267,24 g/mol (anhy) Melting Pt: 300 °C (dec.) Flash Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150760 CAS Number: 961-07-9 EINECS: 213-505-8 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤8.0 % (K.F.) |
HPLC | ≥99.0 % |
UV spectrophotometry | molar absorptivity 13700 +/- 4%, on dry basis |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...