POPSO sesquisodium salt (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid sesquisodium salt)
Supplier: Roth Carl
Synonyms:
POPSO sesquisodium, 3,3'-(Piperazine-1,4-diyl)bis(2-hydroxypropane-1-sulphonic acid), sodium salt(2:3)
6965.1
6965.2
ROTH6965.1EA
0
EUR
ROTH6965.1
ROTH6965.2
POPSO sesquisodium salt (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid sesquisodium salt)
POPSO sesquisodium salt (β,β'-Dihydroxypiperazine-1,4-dipropanesulphonic acid sesquisodium salt)
Formula:
C₂₀H₄₁N₄Na₃O₁₆S₄ MW: 793.8099 g/mol |
MDL Number:
MFCD00070004 CAS Number: 108321-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...