Allyltriphenylphosphonium bromide ≥99%

Supplier: Thermo Fisher Scientific
Warning

A14490.18 A14490.30
A14490.18EA 51.7 EUR
A14490.18 A14490.30
Allyltriphenylphosphonium bromide ≥99%
Allyltriphenylphosphonium bromide

Purity: 99% CAS No.: 1560-54-9 Molecular Formula: C21H20BrP Formula Weight: 383.27 Melting Point: 222-225° Beilstein Reference: 3579053

Formula: C₂₁H₂₀BrP
MW: 383.261 g/mol
Melting Pt: 222…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00011808
CAS Number: 1560-54-9
EINECS: 216-332-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR