4-Bromoanthranilic acid 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Amino-4-bromobenzoic acid

H55829.MD H55829.03 H55829.06
H55829.MDEA 0 EUR
H55829.MD H55829.03 H55829.06
4-Bromoanthranilic acid 97%
4-Bromoanthranilic acid

Purity: 97% CAS No.: 20776-50-5 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 Melting Point: 230-234° MDL No.: MFCD03618454

Formula: C₇H₆BrNO₂
Melting Pt: 230…234 °C
Storage Temperature: Ambient
MDL Number: MFCD03618454
CAS Number: 20776-50-5
EINECS: 244-025-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR