Diallyl trisulphide

Supplier: Cayman Chemical

10012577-25 10012577-100 10012577-250
CAYM10012577-25EA 63.5 EUR
CAYM10012577-25 CAYM10012577-100 CAYM10012577-250
Diallyl trisulphide
Diallyl trisulphide
Formula: C₆H₁₀S₃
MW: 178,34 g/mol
CAS Number: 2050-87-5
EINECS: 218-107-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR