Pentacene 97
Supplier: Thermo Fisher Scientific
Synonyms:
2,3,6,7-Dibenzanthracene
H56310.MD
H56310.03
H56310.MDEA
157
EUR
H56310.MD
H56310.03
Pentacene 97
Pentacene
Formula:
C₂₂H₁₄ MW: 278.35 g/mol Melting Pt: 372…374 °C (subl.) Density: 1.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003710 CAS Number: 135-48-8 EINECS: 205-193-7 Merck Index: 14,7107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...