Myristic acid
Supplier: Cayman Chemical
|
Warning
|
Synonyms:
Tetradecylic acid, Tetradecanoic acid
13351-1
13351-5
13351-10
CAYM13351-1EA
24.4
EUR
CAYM13351-1
CAYM13351-5
CAYM13351-10
Myristic acid
Myristic acid
Formula:
C₁₄H₂₈O₂ MW: 228,38 g/mol Boiling Pt: 319 °C (1013 hPa) Melting Pt: 51…54 °C Density: 0,8622 g/cm³ (20 °C) Flash Pt: >165 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002744 CAS Number: 544-63-8 EINECS: 208-875-2 Merck Index: 12,06416 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...