3-Aminobenzamidoxime

Supplier: Cayman Chemical

11074-500 11074-1 11074-5
CAYM11074-500EA 52.5 EUR
CAYM11074-500 CAYM11074-1 CAYM11074-5
3-Aminobenzamidoxime
3-Aminobenzamidoxime
Formula: C₇H₉N₃O
MW: 151.17 g/mol
MDL Number: MFCD08061150
CAS Number: 100524-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR