4-Methylbenzamidoxime

Supplier: Cayman Chemical

Synonyms: N-Hydroxy-4-methylbenzimidamide

13231-500 13231-1
CAYM13231-500EA 37.9 EUR
CAYM13231-500 CAYM13231-1
4-Methylbenzamidoxime
4-Methylbenzamidoxime
Formula: C₈H₁₀N₂O
MW: 150,18 g/mol
MDL Number: MFCD00019952
CAS Number: 19227-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR