Diethyl chlorophosphate 97

Supplier: Thermo Fisher Scientific
Danger

H33820.14 H33820.22
H33820.14EA 45 EUR
H33820.14 H33820.22
Diethyl chlorophosphate 97
Diethyl chlorophosphate
Formula: C₄H₁₀ClO₃P
MW: 172.55 g/mol
Boiling Pt: 60 °C (2 mmHg)
Density: 1.194
Flash Pt: 61 °C (142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009075
CAS Number: 814-49-3
EINECS: 212-369-4
UN: 3278
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR