Undecanoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
H56380.03
H56380.ME
H56380.03EA
218
EUR
H56380.03
H56380.ME
Undecanoyl chloride ≥98%
Undecanoyl chloride
Formula:
C₁₁H₂₁ClO Boiling Pt: 135…136 °C Flash Pt: 113 °C (235 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00000739 CAS Number: 17746-05-3 EINECS: 241-741-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...