Methyl-4,4,4-trifluoroacetoacetate 95%

Supplier: Thermo Fisher Scientific
Danger

L10140.06 L10140.14
L10140.06EA 64.8 EUR
L10140.06 L10140.14
Methyl-4,4,4-trifluoroacetoacetate 95%
Methyl 4,4,4-trifluoroacetoacetate

Purity: 95% CAS No.: 83643-84-9 Molecular Formula: C5H5F3O3 Formula Weight: 170.09 Flash Point: 40°(104°F) Refractive Index: 1.3720 Boiling Point: 117-118° Beilstein Reference: 1777343 UN No.: 3272 MDL No.: MFCD00041004

Formula: C₅H₅F₃O₃
Boiling Pt: 117…118 °C
Density: 1,340
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041004
CAS Number: 83643-84-9
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR