Benzyloxyacetyl chloride 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(Benzyloxy)acetyl chloride
L14337.03
L14337.06
L14337.14
L14337.03EA
0
EUR
L14337.03
L14337.06
L14337.14
Benzyloxyacetyl chloride 95%
Benzyloxyacetyl chloride
Purity: 95%
CAS No.: 19810-31-2
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₉H₉ClO₂ MW: 184,62 g/mol Boiling Pt: 84…87 °C (0,4 mmHg) Density: 1,189 Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00010768 CAS Number: 19810-31-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...