5-Bromo-2'-deoxyuridine (Broxuridine)

Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

EMR077001 EMR078005 EMR078500
EURLEMR077001EA 0 EUR
EURLEMR077001 EURLEMR078005 EURLEMR078500
5-Bromo-2'-deoxyuridine (Broxuridine)
5-Bromo-2'-deoxyuridine (Broxuridine)
Formula: C₉H₁₁BrN₂O₅
MW: 307,10 g/mol
MDL Number: MFCD00006529
CAS Number: 59-14-3
EINECS: 200-415-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR