Triethyl-2-phosphonobutyrate ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Ethyl 2-(diethoxyphosphoryl)butanoate
L09329.06
L09329.14
L09329.06EA
43.7
EUR
L09329.06
L09329.14
Triethyl-2-phosphonobutyrate ≥97%
Triethyl 2-phosphonobutyrate
Purity: 97%
CAS No.: 17145-91-4
Molecular Formula: C10H21O5P
Formula Weight: 252.25
Flash Point:
Formula:
C₁₀H₂₁O₅P MW: 252.25 g/mol Boiling Pt: 105…106 °C (2 mmHg) Density: 1.064 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041347 CAS Number: 17145-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...