2-(Dimethylamino)ethanethiol hydrochloride 95%

Supplier: Thermo Fisher Scientific
Warning

115810050 115810250 115811000
ACRO115810050EA 61.2 EUR
ACRO115810050 ACRO115810250 ACRO115811000
2-(Dimethylamino)ethanethiol hydrochloride 95%
2-(Dimethylamino)ethanethiol hydrochloride
Formula: C₄H₁₂ClNS
Melting Pt: 154…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012633
CAS Number: 13242-44-9
EINECS: 236-221-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Iodimetric ≥94 %
GC ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7° to -9° (20°C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR