Phthalan ≥96%

Supplier: Thermo Fisher Scientific
Warning

45592.14 45592.22
45592.14EA 0 EUR
45592.14 45592.22
Phthalan ≥96%
Phthalan

Purity: 96% CAS No.: 496-14-0 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 63°(145°F) Physical Form: Liquid Density: 1.094 Refractive Index: 1.5460 Boiling Point: 192° Beilstein Reference: 111932 MDL No.: MFCD00005932

Formula: C₈H₈O
Boiling Pt: 192 °C
Density: 1,094
Flash Pt: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005932
CAS Number: 496-14-0
EINECS: 207-815-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR